accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5585-19-3

IUPAC Name: 5-phenyl-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms: 5-phenyl-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol): 194.27
Molecular Formula: C8H6N2S2
InChi Key: ZTLMHGOWADYAHM-UHFFFAOYSA-N
SMILES: S=C1NN=C(S1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results