accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5585-33-1

IUPAC Name: 2-(morpholin-4-yl)aniline
Synonyms: 2-(morpholin-4-yl)aniline
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: QKWLVAYDAHQMLG-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1N1CCOCC1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)