accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55854-45-0

IUPAC Name: 5-methylthiophene-2-sulfonyl chloride
Synonyms: 5-methylthiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 196.66
Molecular Formula: C5H5ClO2S2
InChi Key: QGGRRHYGHGJEKP-UHFFFAOYSA-N
SMILES: CC1=CC=C(S1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results