accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55854-46-1

IUPAC Name: 5-bromothiophene-2-sulfonyl chloride
Synonyms: 5-bromothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 261.53
Molecular Formula: C4H2BrClO2S2
InChi Key: WGYBIEOLAFYDEC-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(Br)S1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)