accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCl

CAS RN 5586-88-9

IUPAC Name: 1-(4-chlorophenyl)propan-2-one
Synonyms: 1-(4-chlorophenyl)propan-2-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: WEJRYKSUUFKMBC-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results