accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN(E/Z)

CAS RN 55873-21-7

IUPAC Name: 4-{[(4-butoxyphenyl)imino]methyl}benzonitrile
Synonyms: 4-{[(4-butoxyphenyl)imino]methyl}benzonitrile
Molecular Weight (g/mol): 278.36
Molecular Formula: C18H18N2O
InChi Key: NFABQUNUXAXGBJ-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)N=CC1=CC=C(C=C1)C#N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results