accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOOONHNH(R)CH3CH3H3C

CAS RN 55878-47-2

IUPAC Name: (2R)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2R)-6-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 380.44
Molecular Formula: C19H28N2O6
InChi Key: BDHUTRNYBGWPBL-OAHLLOKOSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)