accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHVOHH3CCH3OHCH3CH3O

CAS RN 5588-84-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 247.23
Molecular Formula: C9H24O4V
InChi Key: JOUSPCDMLWUHSO-UHFFFAOYSA-N
SMILES: O=[V].CC(C)O.CC(C)O.CC(C)O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (5)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (7)
  • (2)
  • (3)