accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClBr

CAS RN 5589-96-8

IUPAC Name: 2-bromo-2-chloroacetic acid
Synonyms: bromochloroacetic acid keratin
Molecular Weight (g/mol): 173.39
Molecular Formula: C2H2BrClO2
InChi Key: GEHJBWKLJVFKPS-UHFFFAOYNA-N
SMILES: OC(=O)C(Cl)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results