accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNHNOONOO

CAS RN 55907-61-4

IUPAC Name: hydrogen (2,4-dinitrophenyl)hydrazine chloride
Synonyms: hydrogen 2,4-dinitrophenylhydrazine chloride
Molecular Weight (g/mol): 234.6
Molecular Formula: C6H7ClN4O4
InChi Key: PUYFAZQODQZOFK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)