accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55912-20-4

IUPAC Name: (4-chloro-3-nitrophenyl)methanol
Synonyms: (4-chloro-3-nitrophenyl)methanol
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: QLLRQJDSYJIXTN-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)