accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55934-00-4

IUPAC Name: 3,4-dichloropyridine
Synonyms: 3,4-dichloropyridine
Molecular Weight (g/mol): 147.99
Molecular Formula: C5H3Cl2N
InChi Key: ZMPYQKNNUHPTLT-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=NC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results