accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 55963-78-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C10H11NaO4S)n
InChi Key:
SMILES:
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)