accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOCH3ONClClCl

CAS RN 5598-13-0

IUPAC Name: O,O-dimethyl O-3,5,6-trichloropyridin-2-yl phosphorothioate
Synonyms: tumar chlorpyrifos methyl reldan 50 EC caswell no. 179AA chlorpyrifos-methyl methyl dursban methyl chlorpyrifos chloropyriphos-methyl epa pesticide chemical code 059102 zertell
Molecular Weight (g/mol): 322.52
Molecular Formula: C7H7Cl3NO3PS
InChi Key: HRBKVYFZANMGRE-UHFFFAOYSA-N
SMILES: COP(=S)(OC)OC1=NC(Cl)=C(Cl)C=C1Cl
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)