accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56-24-6

IUPAC Name: trimethylstannanol
Synonyms: trimethyltin hydroxide
Molecular Weight (g/mol): 180.82
Molecular Formula: C3H10OSn
InChi Key: OJZNYUDKNVNEMV-UHFFFAOYSA-M
SMILES: C[Sn](C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)