accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)(S)(S)(S)(S)(S)HCH3HCH3OOCH3OOH

CAS RN 56-47-3

IUPAC Name: 2-[(1S,3aS,3bS,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms: desoxycorticosterone acetate
Molecular Weight (g/mol): 372.51
Molecular Formula: C23H32O4
InChi Key: VPGRYOFKCNULNK-ZIUYWEQENA-N
SMILES: [H][C@@]12CC[C@H](C(=O)COC(C)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])CCC2=CC(=O)CC[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results