accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 56-49-5

IUPAC Name: {1H,2H-benzo[j]aceanthrylen-3-yl}methane
Synonyms: methylcholanthrene 3MC 3-mca 3-mch 3-methylbenz(j)aceanthrene MCA 3-MeCA 3-methylchloanthrene 20-methylcholanthrene ccris 386
Molecular Weight (g/mol): 268.36
Molecular Formula: C21H16
InChi Key: PPQNQXQZIWHJRB-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C3C(C=CC4=CC=CC=C34)=C3CCC1=C23
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results