accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONH2OHHO

CAS RN 56-69-9

IUPAC Name: 2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Synonyms: hydroxytryptophan|DL-5-hydroxytryptophan|5-hydroxy-DL-tryptophan|DL-5-htp|5-hydroxytryptophan DL-form
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: LDCYZAJDBXYCGN-UHFFFAOYSA-N
SMILES: NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results