accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 560090-29-1

IUPAC Name: N-(2-ethylhexyl)-4-methoxybenzamide
Synonyms: N-(2-ethylhexyl)-4-methoxybenzamide
Molecular Weight (g/mol): 263.38
Molecular Formula: C16H25NO2
InChi Key: OAGPSBYQMCQNOF-UHFFFAOYNA-N
SMILES: CCCCC(CC)CNC(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results