CAS RN 56011-12-2
IUPAC Name:
2,2-diethoxypropanenitrile
Synonyms:
2,2-diethoxypropanenitrile
Molecular Weight (g/mol):
143.19
Molecular Formula:
C7H13NO2
InChi Key:
QPFCLGADWIWYFW-UHFFFAOYSA-N
SMILES:
CCOC(C)(OCC)C#N
1
–
1
of
1
results
1
2,2-Diethoxypropionitrile 98.0+%, TCI America™
CAS: 56011-12-2 Molecular Formula: C7H13NO2 Molecular Weight (g/mol): 143.186 MDL Number: MFCD00209573 InChI Key: QPFCLGADWIWYFW-UHFFFAOYSA-N PubChem CID: 566609 IUPAC Name: 2,2-diethoxypropanenitrile SMILES: CCOC(C)(C#N)OCC