accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(R)O(R)ClO(S)Cl(S)HO(S)HO(R)HO(R)OH(R)Cl

CAS RN 56038-13-2

IUPAC Name: (2R,3R,4R,5R,6R)-2-{[(2R,3S,4S,5S)-2,5-bis(chloromethyl)-3,4-dihydroxyoxolan-2-yl]oxy}-5-chloro-6-(hydroxymethyl)oxane-3,4-diol
Synonyms: splenda sucralose trichlorogalactosucrose trichlorogalacto-sucrose sucralose (substance) trichlorosucrose
Molecular Weight (g/mol): 397.63
Molecular Formula: C12H19Cl3O8
InChi Key: BAQAVOSOZGMPRM-QBMZZYIRSA-N
SMILES: OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl
Percent Purity 
  • (1)
  • (3)
  • (7)
  • (3)
Physical Form 
  • (3)
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (7)
  • (3)
  • (3)
  • (7)
  • (1)
  • (3)
  • (7)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)