accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5606-24-6

IUPAC Name: N2-butyl-1,3,5-triazine-2,4,6-triamine
Synonyms: N2-butyl-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol): 182.23
Molecular Formula: C7H14N6
InChi Key: CVKGSDYWCFQOKU-UHFFFAOYSA-N
SMILES: CCCCNC1=NC(N)=NC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results