CAS RN 5610-49-1
IUPAC Name:
1-butylpiperazine
Synonyms:
1-butylpiperazine
Molecular Weight (g/mol):
142.25
Molecular Formula:
C8H18N2
InChi Key:
YKSVXVKIYYQWBB-UHFFFAOYSA-N
SMILES:
CCCCN1CCNCC1
1
–
1
of
1
results
1
1-Butylpiperazine 98.0+%, TCI America™
CAS: 5610-49-1 Molecular Formula: C8H18N2 Molecular Weight (g/mol): 142.246 MDL Number: MFCD00039702 InChI Key: YKSVXVKIYYQWBB-UHFFFAOYSA-N PubChem CID: 424322 IUPAC Name: 1-butylpiperazine SMILES: CCCCN1CCNCC1