accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHOH3CCH3CH3OHCH3

CAS RN 5613-46-7

IUPAC Name: 4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]-2,6-dimethylphenol
Synonyms: 4-[2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]-2,6-dimethylphenol
Molecular Weight (g/mol): 284.4
Molecular Formula: C19H24O2
InChi Key: ODJUOZPKKHIEOZ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1O)C(C)(C)C1=CC(C)=C(O)C(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)