accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOONOO

CAS RN 56136-84-6

IUPAC Name: 1-(6-nitro-2H-1,3-benzodioxol-5-yl)ethan-1-one
Synonyms: 1-(6-nitro-2H-1,3-benzodioxol-5-yl)ethanone
Molecular Weight (g/mol): 209.16
Molecular Formula: C9H7NO5
InChi Key: BQONDGIXVHVIIR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC2=C(OCO2)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results