accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5619-04-5

IUPAC Name: hydrogen methyl 2-amino-3-hydroxypropanoate chloride
Synonyms: hydrogen methyl 2-amino-3-hydroxypropanoate chloride
Molecular Weight (g/mol): 155.58
Molecular Formula: C4H10ClNO3
InChi Key: NDBQJIBNNUJNHA-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(N)CO
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)