accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 562-90-3

IUPAC Name: tris(acetyloxy)silyl acetate
Synonyms: silicon tetraacetate
Molecular Weight (g/mol): 264.26
Molecular Formula: C8H12O8Si
InChi Key: YZVRVDPMGYFCGL-UHFFFAOYSA-N
SMILES: CC(=O)O[Si](OC(C)=O)(OC(C)=O)OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)