accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56211-40-6

IUPAC Name: 1-({4-[(3-methylphenyl)amino]pyridin-3-yl}sulfonyl)-3-(propan-2-yl)urea
Synonyms: torasemide luprac torasemida torsemide demadex torasemida [inn-spanish] torsemide preparation torsemide (substance) torasemide (product) torasemidum
Molecular Weight (g/mol): 348.42
Molecular Formula: C16H20N4O3S
InChi Key: NGBFQHCMQULJNZ-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)NS(=O)(=O)C1=CN=CC=C1NC1=CC=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results