accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5622-78-6

IUPAC Name: 2,4,5-trimethylpyridin-3-ol
Synonyms: 2,4,5-trimethylpyridin-3-ol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: OIXVDFZUALXJNM-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C(O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results