accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56239-26-0

IUPAC Name: hydrogen 4-aminocyclohexan-1-ol chloride
Synonyms: hydrogen 4-aminocyclohexan-1-ol chloride
Molecular Weight (g/mol): 151.63
Molecular Formula: C6H14ClNO
InChi Key: RKTQEVMZBCBOSB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results