accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONO

CAS RN 5625-90-1

IUPAC Name: 4-[(morpholin-4-yl)methyl]morpholine
Synonyms: 4,4-methylenedimorpholine
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: MIFZZKZNMWTHJK-UHFFFAOYSA-N
SMILES: C(N1CCOCC1)N1CCOCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)