accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSN

CAS RN 56278-50-3

IUPAC Name: 2-(1,3-benzothiazol-2-yl)acetonitrile
Synonyms: 2-(1,3-benzothiazol-2-yl)acetonitrile
Molecular Weight (g/mol): 174.22
Molecular Formula: C9H6N2S
InChi Key: ZMZSYUSDGRJZNT-UHFFFAOYSA-N
SMILES: N#CCC1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results