accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClH2C

CAS RN 563-52-0

IUPAC Name: 3-chlorobut-1-ene
Synonyms: 3-chloro-1-butene
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: VZGLVCFVUREVDP-UHFFFAOYNA-N
SMILES: CC(Cl)C=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)