accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 563-96-2

IUPAC Name: 2,2-dihydroxyacetate
Synonyms: 2,2-dihydroxyacetate
Molecular Weight (g/mol): 91.04
Molecular Formula: C2H3O4
InChi Key: GOCCREQJUBABAL-UHFFFAOYSA-M
SMILES: OC(O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)