accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56309-56-9

IUPAC Name: 1-isocyanato-2-(propan-2-yl)benzene
Synonyms: 1-isocyanato-2-isopropylbenzene
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: MWBSQVPLYJZPTG-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1N=C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results