accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56341-36-7

CH3NCH3N

CAS RN 56341-36-7

IUPAC Name: 1,5-dimethyl-1H-pyrrole-2-carbonitrile
Synonyms: 1,5-dimethylpyrrole-2-carbonitrile
Molecular Weight (g/mol): 120.16
Molecular Formula: C7H8N2
InChi Key: DRXOPQFEWDRGKT-UHFFFAOYSA-N
SMILES: CN1C(C)=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results