accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56363-84-9

IUPAC Name: 2-chloro-4-hydroxybenzoic acid
Synonyms: 2-chloro-4-hydroxybenzoic acid
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: WIPYZRZPNMUSER-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C=C1Cl
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)