accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH2H2N

CAS RN 5637-42-3

IUPAC Name: N''-(4-cyanophenyl)guanidine
Synonyms: N''-(4-cyanophenyl)guanidine
Molecular Weight (g/mol): 160.18
Molecular Formula: C8H8N4
InChi Key: BTDGLZSKNFJBER-UHFFFAOYSA-N
SMILES: NC(N)=NC1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results