accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHN

CAS RN 5638-76-6

IUPAC Name: methyl[2-(pyridin-2-yl)ethyl]amine
Synonyms: betahistine betahistine methanesulphonate methanesulfonate, betahistine betahistin-ratiopharm by vertin betavert by-vertin betaistina extovyl betahistine mesylate
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: UUQMNUMQCIQDMZ-UHFFFAOYSA-N
SMILES: CNCCC1=CC=CC=N1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results