accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56413-96-8

IUPAC Name: 5-amino-6-chloropyrazine-2,3-dicarbonitrile
Synonyms: 5-amino-6-chloropyrazine-2,3-dicarbonitrile
Molecular Weight (g/mol): 179.57
Molecular Formula: C6H2ClN5
InChi Key: CDKQWBZSAPRZNS-UHFFFAOYSA-N
SMILES: NC1=NC(C#N)=C(N=C1Cl)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results