accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSNH3CS

CAS RN 56421-62-6

IUPAC Name: ethyl 4-methyl-2-(thiophen-2-yl)-1,3-thiazole-5-carboxylate
Synonyms: ethyl 4-methyl-2-(thiophen-2-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 253.33
Molecular Formula: C11H11NO2S2
InChi Key: NRTAQEAHFFIBFX-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(S1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results