accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56442-17-2

IUPAC Name: 4-[(4-fluorophenyl)methoxy]benzaldehyde
Synonyms: 4-[(4-fluorophenyl)methoxy]benzaldehyde
Molecular Weight (g/mol): 230.24
Molecular Formula: C14H11FO2
InChi Key: QGULWBQOCMQNFD-UHFFFAOYSA-N
SMILES: FC1=CC=C(COC2=CC=C(C=O)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results