accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 564483-19-8

IUPAC Name: di-tert-butyl[2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: di-tert-butyl({2',4',6'-triisopropyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 424.65
Molecular Formula: C29H45P
InChi Key: SACNIGZYDTUHKB-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)C1=CC=CC=C1P(C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)