accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56456-47-4

IUPAC Name: (2,4-difluorophenyl)methanol
Synonyms: (2,4-difluorophenyl)methanol
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: NIJZBWHOHNWJBX-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results