accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56456-50-9

IUPAC Name: (2-chloro-6-fluorophenyl)methanol
Synonyms: (2-chloro-6-fluorophenyl)methanol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: ZZFCUESFXBCRSC-UHFFFAOYSA-N
SMILES: OCC1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results