accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3HO

CAS RN 565-60-6

IUPAC Name: 3-methylpentan-2-ol
Synonyms: 3-methyl-2-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: ZXNBBWHRUSXUFZ-UHFFFAOYNA-N
SMILES: CCC(C)C(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results