accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O(Z)

CAS RN 565-62-8

IUPAC Name: (3Z)-3-methylpent-3-en-2-one
Synonyms: 3-methyl-3-penten-2-one 3-methylpent-3-EN-2-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ZAMCMCQRTZKGDX-PLNGDYQASA-N
SMILES: C\C=C(\C)C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)