accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHO(Z)

CAS RN 565-63-9

IUPAC Name: (2Z)-2-methylbut-2-enoic acid
Synonyms: angelic acid 2-methylisocrotonic acid angelicasaeure α-methyl isocrotonic acid acide angelique angelikasaeure cis-2-dimethylcrotonic acid acido angelico α-methylisocrotonic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: UIERETOOQGIECD-ARJAWSKDSA-N
SMILES: C\C=C(\C)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)