accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 565-73-1

IUPAC Name: sodium carbamoylformate
Synonyms: sodium oxamate
Molecular Weight (g/mol): 111.03
Molecular Formula: C2H2NNaO3
InChi Key: RQVZIJIQDCGIKI-UHFFFAOYSA-M
SMILES: [Na+].NC(=O)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)