accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 56538-57-9

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-2-hydroxyacetic acid
Synonyms: {[(benzyloxy)carbonyl]amino}(hydroxy)acetic acid
Molecular Weight (g/mol): 225.2
Molecular Formula: C10H11NO5
InChi Key: JQEZLSUFDXSIEK-UHFFFAOYNA-N
SMILES: OC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)